missing translation for 'onlineSavingsMsg'
Learn More

3M™ Chemical Spill Kit

Product Code. 17139350
Change view
Click to view available options
Includes:
P110 Sheets (10), P300 Pillow, Disposal Bag w/ Ties
P110 Sheets (100), P300 Pillows (12), P200 Mini-booms (8), Disposal Bags w/ Ties (5)
Unit Size:
1 set
5 sets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17139350 P110 Sheets (10), P300 Pillow, Disposal Bag w/ Ties 5 sets
17129350 P110 Sheets (100), P300 Pillows (12), P200 Mini-booms (8), Disposal Bags w/ Ties (5) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17139350 Supplier 3M™ Supplier No. 7100090374
  • ONLINE EXCLUSIVE PRICE
    567.00 €
    350.96 € / 5 sets
    Save 216.04 €
    38% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensures that everything is available, and ready should a spill occur. Cleaning up hazardous chemical spills quickly in an emergency situation promotes on-site safety.

  • Ideal for rapid response to chemical spills: fast and effective containment and clean-up
  • Cleaning up hazardous chemical spills, drips, and leaks
  • Suitable for use with most chemicals
  • Promotes on-site safety
  • Easy to dispose of
TRUSTED_SUSTAINABILITY

Specifications

Type Chemical Spill Response Kit
Format Kit
For Use With (Application) Rapid Response to Chemical Spills
Includes P110 Sheets (10), P300 Pillow, Disposal Bag w/ Ties
Model SK 5
Packaging Quantity 5 Pk.
Dimensions (L x W) 400 x 70 mm
Length (Metric) 400 mm
Width (Metric) 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.