missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chemical Test Kit

Product Code. 11642360
Change view
Click to view available options
For Use With (Application):
Acidity Determination
Ammonia Determination in Fresh Water
Ammonia Determination in Seawater
Chloride Determination
Phosphate Determination
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
11642360 Chloride Determination Each
11612370 Acidity Determination Each
10480684 Ammonia Determination in Fresh Water Each
11692370 Ammonia Determination in Seawater Each
10634155 Phosphate Determination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11642360 Supplier Hanna Instruments™ Supplier No. HI3815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted

Specifications

Analytes Mercuric Nitrate
For Use With (Application) Chloride Determination
For Use With (Equipment) Water samples
Includes 1 (15mL) dropper bottle diphenylcarbazone indicator, 1 (30mL) dropper bottle nitric acid solution, 120mL bottle mercuric nitrate solution, 50mL calibrated vessel, 10mL calibrated vessel, calibrated syringe with tip
No. of Tests 110
Quantity 1 Kit
Range 0 to 100mg/L, 0 to 1000mg/L
Test Method Titration
Type Chloride Test Kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1 SET CHLORIDE TEST KIT HI4815
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use powder to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, mercury(II)nitrate, nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.