Learn More
Hanna Instruments™ Chemical Test Kit
Product Code. 11692370
Change view
Click to view available options
For Use With (Application):
Acidity Determination
Ammonia Determination in Fresh Water
Ammonia Determination in Seawater
Chloride Determination
Phosphate Determination
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11692370 Ammonia Determination in Seawater Each
11642360 Chloride Determination Each
11612370 Acidity Determination Each
10480684 Ammonia Determination in Fresh Water Each
10634155 Phosphate Determination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11692370 Supplier Hanna Instruments™ Supplier No. HI3826
  • ONLINE EXCLUSIVE PRICE
    213.59 €
    83.97 € / Pack of 1
    Save 129.62 €
    60% Off
View all promotions
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
For Use With (Application) Ammonia Determination in Seawater
For Use With (Equipment) Water samples
Includes 20mL plastic beaker, color comparison cube, 20mL dropper bottle ammonia reagent 1 (for seawater), and 20mL dropper bottle Nessler reagent
No. of Tests 25
Quantity 1 Kit
Range 0.0 to 2.5mg/L
Test Method Titration
Type Chemical Test Kit