missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ CRF1 Receptor Membrane Preparation

Product Code. 17374999
Encompass
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
1 ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17374999 1 Vial 1 ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17374999 Supplier DiscoverX Supplier No. HTS023M
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The ChemiScreen™ CRF1 Receptor Membrane Preparation allows measurement of ligand binding to CRHR1 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.

The ChemiScreen™ CRF1 Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CRHR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.

Specifications

Assay Entry CRF1
For Use With (Application) Membrane Preps
Quantity 1 Vial
Target Entry CRHR1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.