missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves

Product Code. 12629711
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12629711 10 1 pair
12649711 7 1 pair
12639711 11 1 pair
12659711 8 1 pair
12669711 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12629711 Supplier Ansell™ Supplier No. 38520100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. ChemTekTM gloves are surprisingly flexible and provide excellent dexterity.

  • Smooth finish in black
  • Easy donning and good grip
  • Maximum protection from hazardous chemicals
  • Surprisingly flexible and provide excellent dexterity
  • Designed for comfort with a natural, curved ergonomic shape and soft feel
  • Absence of latex proteins means there's no risk of latex allergies
  • Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace
TRUSTED_SUSTAINABILITY

Specifications

Back Material Butyl
Cuff Material Butyl
Applications Industrial
Coating Material Butyl Polymer
Color Black
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Butyl Polymer
Product Type Immersion Gloves
Size 10
Thickness (English) 20 mil
Glove Cuff Color Black
Glove Cuff Style Beaded/Rolled
Hem Color Black
Inspected for Medical Use Yes
Packaging Type Individually wrapped bags with insert
Quantity 1 Pair
Resistant To Highly chemical resistance
Series 38-520
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.