missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves
Description
- Smooth finish in black
- Easy donning and good grip
- Maximum protection from hazardous chemicals
- Surprisingly flexible and provide excellent dexterity
- Designed for comfort with a natural, curved ergonomic shape and soft feel
- Absence of latex proteins means there's no risk of latex allergies
- Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace
Specifications
Specifications
| Back Material | Butyl |
| Cuff Material | Butyl |
| Applications | Industrial |
| Coating Material | Butyl Polymer |
| Color | Black |
| Laboratory/Cleanroom Use | Yes |
| Latex Free | Yes |
| Material | Butyl Polymer |
| Product Type | Immersion Gloves |
| Size | 7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?