missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chenodeoxycholic acid, 98%

Product Code. 10266950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10266950 5 g Glass bottle 5g
10521291 25 g Glass bottle 25g
10368613 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10266950 Supplier Thermo Scientific Chemicals Supplier No. 228770050
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Chenodeoxycholic acid, 98%, ACROS Organics, Quantity: 5g, Packaging: Glass bottle, White, Melting Point: 162 deg.C to 168 deg.C, Molecular Weight: 391.572, Percent Purity: >-97.5%, Assay Percent Range: 97+%, CAS: 474-25-9, ChEBI: CHEBI:36234, EINECS Number: 207-481-8, Formula Weight: 392.57

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 474-25-9
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00064142
InChI Key RUDATBOHQWOJDD-BSWAIDMHSA-N
Synonym chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
PubChem CID 9548590
ChEBI CHEBI:36234
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifikationer

Melting Point 162°C to 168°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Specific Rotation +10° to +13° (20°C, 589nm) (c=1, EtOH)
Formula Weight 392.57
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Chenodeoxycholic acid, 98%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.