missing translation for 'onlineSavingsMsg'
Learn More

Chenodeoxycholic Acid, MedChemExpress

Product Code. 16437956
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437956 100 mg 100mg
16441956 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437956 Supplier Medchem Express Supplier No. HY76847
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.

Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.

Specifications

Chemical Name or Material Chenodeoxycholic Acid
CAS 474-25-9
Color White
Purity Grade Notes Research
Molecular Formula C24H40O4
Quantity 100 mg
Synonym CDCA
Solubility Information DMSO : ≥ 50 mg/mL (127.37 mM)
SMILES O=C(O)CC[C@@H](C)[C@]1([H])[C@]2(C)[C@](CC1)([H])[C@]3([H])[C@H](O)C[C@@](C4)([H])[C@@](CC[C@H]4O)(C)[C@]([H])3CC2
Molecular Weight (g/mol) 392.57
Formula Weight 392.57
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.