missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594

Product Code. 10206452
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10206452 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10206452 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' A21468
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Secondary Antibody

This Chicken anti-goat antibody reacts with IgG heavy chains and all classes of immunoglobulin light chains from goat. Chicken secondary antibodies have gained popularity because they demonstrate a lower level of nonspecific binding. Chicken antibodies lack a classic 'Fc' domain and will not bind to protein A or protein G, nor will they bind to mammalian IgG Fc receptors. Product will be shipped at Room Temperature.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat IgG (H+L) Cross-Adsorbed
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 594
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Chicken
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.