missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Human IgG Fc Secondary Antibody, Biotin

Product Code. 11505170
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11505170 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11505170 Supplier Invitrogen™ Supplier No. SA172048
  • ONLINE EXCLUSIVE PRICE
    370.00 €
    329.30 € / 500µg
    Save 40.70 €
    11% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Secondary Antibody

SA1-72048 detects Human IgG Fc fragment in human samples. SA1-72048 has been successfully used in ELISA and Western blot procedures. The SA1-72048 immunogen is from purified IgG from normal human serum. Store at -20°C for long term storage. Avoid freeze/thaw cycles.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG Fc
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.1% chloroacetamide
Host Species Chicken
Immunogen Purified IgG from normal human serum.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.