missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, AP

Product Code. 15201648
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15201648 500 μg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15201648 Supplier Invitrogen™ Supplier No. A18736
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    166.43 € / 0.5mg
    Save 20.57 €
    11% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Secondary Antibody

Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody for ELISA, IHC, Western Blot

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG (H+L) Cross-Adsorbed
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation Secondary antibody with 30mM triethanolamine, 1% BSA, 5mM MgCl2, 0.1mM ZnCl2 and 0.05% sodium azide; pH 7.2
Host Species Chicken
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.