missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicoric acid, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Description
Chicoric acid inhibits cell viability and induces mitochondria-dependent apoptosis in 3T3-L1 preadipocytes through ROS-mediated PI3K/Akt and MAPK signaling pathways. Chicoric acid increases glucose uptake, improves insulin resistance, and attenuates glucosamine-induced inflammation. Chicoric acid has antidiabetic properties and antioxidant, anti-inflammatory effects.
Specifications
Specifications
| Chemical Name or Material | Chicoric acid |
| CAS | 6537-80-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18O12 |
| Quantity | 50 mg |
| Synonym | Cichoric acid Dicaffeoyltartaric acid |
| Solubility Information | H2O : 100 mg/mL (210.81 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.11 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@H](OC(/C=C/C1=CC=C(O)C(O)=C1)=O)[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C(O)=O |
| Molecular Weight (g/mol) | 474.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction