Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Marca: Thermo Scientific™ 3330292
Code : 04
Detalles adicionales : Weight : 10.00000kg
Descripción
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Especificaciones
Tubing, Insulation | |
Tubing | |
6 mm ID per meter |
ThermoTemp, Polar, VersaCool | |
Insulated flexible SS tubing with M16x1 threaded fittings 0.5 meter long |