Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
47.80 € - 1350.00 €
Product Code | Brand | Description | For Use With (Equipment) | Size | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Size | Includes | Price | Quantity & Availability | ||
15176445
|
Thermo Scientific™
611000000012 |
25 ft. Plastic Plumbing Package | Merlin™ Chillers with a PD1 or PD2 pump | 1/2 in. NPT | 25 ft. of 1/2 in. ID flexible tubing, 25 ft. of tubing insulation, and hose clamp |
343.00 €
Each |
In Stock
Log in to see stock. |
||
17108165
|
Thermo Scientific™
611000000107 |
10m Plumbing Package | ThermoFlex™ 3500 to 5000 with P3, P4 pumps | 3/4 in. NPT | 32.8 ft. (10 meters) of 5/8 in. ID Polybraid hose (use with 3/4 in. hose barbs) , insulation and clamps |
405.00 €
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
11957514
|
Thermo Scientific™
611000000100 |
50 ft. Standard Temperature Plumbing Package | ThermoFlex™ 900 to 10000 with P1, P2 or T1 pumps | 1/2 in. NPT | 50 ft. of 5/8 in. ID Polybraid hose comes with 1/2 in. hose barbs, insulation and clamps |
362.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
10430463
|
Thermo Scientific™
3330293 |
Tubing, Insulation | ThermoTemp, Polar, VersaCool | 6 mm ID per meter | Insulated flexible SS tubing with M16x1 threaded fittings 1.0 meter long |
483.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
10505872
|
Thermo Scientific™
3330578 |
Tubing, Insulation | ThermoTemp, Polar | 6 mm ID per meter | Insulated flexible SS tubing with M16x1 threaded fittings 1.0 meter long |
1130.00 €
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||
10771993
|
Thermo Scientific™
3330579 |
Tubing, Insulation | ThermoTemp, Polar | 6 mm ID per meter | Insulated flexible SS tubing with M16x1 threaded fittings 1.5 meter long |
1350.00 €
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||
10719181
|
Thermo Scientific™
3330292 |
Tubing, Insulation | ThermoTemp, Polar, VersaCool | 6 mm ID per meter | Insulated flexible SS tubing with M16x1 threaded fittings 0.5 meter long |
407.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
10111031
|
Thermo Scientific™
3330294 |
Tubing, Insulation | ThermoTemp, Polar, VersaCool | 6 mm ID per meter | Insulated flexible SS tubing with M16x1 threaded fittings 1.5 meter long |
613.00 €
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
10258621
|
Thermo Scientific™
0820172 |
Tubing, Perbunan | ThermoTemp, Polar | 8 mm ID per meter | PERBUNAN Tubing, 8mm ID custom length |
47.80 €
1m |
In Stock
Log in to see stock. |
||
15865038
|
Thermo Scientific™
598000000011 |
Hose Barbs | ThermoFlex™ 900 to 10000 with P1, P2 or T1 pumps | 1/2 to 3/8 in. NPT | Two 1/2 in. MPT to 3/8 in. ID brass hose barbs |
96.60 €
Pack of 2 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps