missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chiller Plumbing Kits and Accessories

Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.

Brand:  Thermo Scientific™ 3330579

Code : 04

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 10771993

  • 1350.00 € / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.

Features:

  • Include flexible tubing, tubing insulation, and hose clamps
Specifications

Specifications

Tubing, Insulation
-90°C to +150°C
Insulated flexible SS tubing with M16x1 threaded fittings 1.5 meter long
ThermoTemp, Polar
Tubing
6 mm ID per meter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers