Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Brand:  Thermo Scientific™ 611000000100
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11957514
  • 362.00 € / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
  • Include flexible tubing, tubing insulation, and hose clamps
Specifications
Specifications
50 ft. Standard Temperature Plumbing Package
Tubing Kit
1/2 in. NPT
ThermoFlex™ 900 to 10000 with P1, P2 or T1 pumps
50 ft. of 5/8 in. ID Polybraid hose comes with 1/2 in. hose barbs, insulation and clamps