missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CHIR 99021

Product Code. 30021991
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30021991 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021991 Supplier R&D Systems™ Supplier No. TB4423GMP/10
  • ONLINE EXCLUSIVE PRICE
    1386.31 €
    1230.60 € / 10mg
    Save 155.71 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CHIR 99021 synthesized to cGMP guidelines

CHIR 99021 synthesized to cGMP guidelines. Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

For more information about how CHIR 99021 may be used, please see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts, Reprogramming MEFs into Skeletal Muscle Progenitors, Highly Efficient Generation of CiPSCs from MEFs, Generation of Kidney Organoids from hPSCs, Culturing Transcriptomically Distinct Pluripotent mESCs .

Research use only CHIR 99021 also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-[[2-[[4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile
CAS 252917-06-9
Quantity 10 mg
Product Line Tocris
Target GSK-3 Inhibitors
Molecular Formula C22H18Cl2N8
Purity 99%
Regulatory Status GMP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.