missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CHIR 99021

Product Code. 18085102
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085102 10 mg 10mg
30001469 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085102 Supplier R&D Systems™ Supplier No. 4423/10
  • ONLINE EXCLUSIVE PRICE
    363.21 €
    325.50 € / 10mg
    Save 37.71 €
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly selective GSK-3 inhibitor

CHIR 99021 is a potent and highly selective inhibitor of glycogen synthase kinase 3 (GSK-3) (IC50 values are 6.7 and 10 nM for GSK-3β and GSK-3α respectively). Acts as Wnt activator. Exhibits >500-fold selectivity for GSK-3 over closely related kinases; also displays >800-fold selectivity against 45 additional enzymes and receptors. In combination with the irreversible inhibitor of LSD1 (Cat. No. 3852), CHIR 99021 enables reprogramming of mouse embryonic fibroblasts, transduced by Oct4 and Klf4 only, into iPSCs. Enhances mouse and human ESC self-renewal when used in combination with PD 0325901 (Cat. No. 4192). It promotes differentiation of hPSCs in cardiomyocytes and allows formation of extended pluripotent stem (EPS) cells in combination with (S)-(+)-Dimethindene maleate (Cat. No. 1425), tetracycline antibiotic (Cat. No. 3268) and human leukemia inhibitory factor. CHIR 99021 synthesized to cGMP guidelines also available. For more information on how to use CHIR 99021, see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252917-06-9
Molecular Formula C22H18Cl2N8

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 465.34
Purity >99%
Molecular Weight (g/mol) 465.34
Chemical Name or Material CHIR 99021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.