missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CHIR99021

Product Code. p-7217591
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17190012

Brand: STEMCELL Technologies 72054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WNT pathway activator; Inhibits GSK3

  • CHIR99021 is an aminopyrimidine derivative that is an extremely potent glycogen synthase kinase (GSK) 3 inhibitor, inhibiting both GSK3β (IC50 = 6.7 nM) and GSK3α (IC50 = 10 nM)
  • GSK3 is a serine/threonine kinase that is a key inhibitor of the WNT pathway; therefore CHIR99021 functions as a WNT activator
  • It shows little activity against a large panel of kinases including CDK2 and other serine/threonine kinases such as MAPK and PKB

Specifications

CAS 252917-06-9
Synonym CT 99021
Quantity 10 mg
Molecular Formula C22H18Cl2N8
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.