missing translation for 'onlineSavingsMsg'
Learn More

Chitin, Practical Grade, TRC

Product Code. 30009823
Change view
Click to view available options
Quantity:
250 mg
1 g
100 g
Unit Size:
100g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009823 1 g 1g
30010606 100 g 100g
30007177 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009823 Supplier Toronto Research Chemicals Supplier No. TRCC3008001G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1398-61-4
Molecular Formula C24 H39 N3 O15 X2
Molecular Weight (g/mol) 609.58
IUPAC Name N-[(2R,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES [X]O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H]([X])[C@H](O)[C@H]3NC(=O)C)[C@H](O)[C@H]2NC(=O)C)[C@H](O)[C@H]1NC(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8-/m1/s1
Formula Weight 609.2381
Chemical Name or Material Chitin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.