missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chitosan, molecular weight: 100,000-300,000

Product Code. 10224991 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
50g
This item is not returnable. View return policy

Product Code. 10224991

Brand: Thermo Scientific Chemicals undefined

Only null left
Add to basket
This item is not returnable. View return policy

Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Chitosan is a naturally derived polymer formed from the deacetylation of chitin
  • It is an alkaline polysaccharide with moisturizing and adsorption properties
  • Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups

Application

  • Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.464
MDL Number MFCD00161512
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
PubChem CID 71853
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O

Specifications

Color Beige-Yellow to White
Physical Form Powder or Granules
Ash 2% max.
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% max. (K.F.)
Chemical Name or Material Chitosan, Molecular Weight: 100, 000 to 300, 000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.