missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chitosan, molecular weight: 100,000-300,000

Product Code. 10732954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10732954 500 g Plastic bottle 500g
10416393 50 g Plastic bottle 50g
10224991 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10732954 Supplier Thermo Scientific Chemicals Supplier No. 349055000
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Chitosan is a naturally derived polymer formed from the deacetylation of chitin
  • It is an alkaline polysaccharide with moisturizing and adsorption properties
  • Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups

Application

  • Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.464
MDL Number MFCD00161512
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
PubChem CID 71853
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O

Specifications

Color Beige-Yellow to White
Physical Form Powder or Granules
Ash 2% max.
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% max. (K.F.)
Chemical Name or Material Chitosan, Molecular Weight: 100, 000 to 300, 000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.