missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chitosan, molecular weight: 600,000-800,000

Product Code. 10636695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636695 50 g Plastic bottle 50g
10421610 100 g Plastic bottle 100g
10616290 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636695 Supplier Thermo Scientific Chemicals Supplier No. 428850500
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Chitosan is a naturally derived polymer formed from the deacetylation of chitin
• It is an alkaline polysaccharide with moisturizing and adsorption properties
• Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups

Application
• Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.464
MDL Number MFCD00161512
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
PubChem CID 71853
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O

Spezifikation

Color White
Physical Form Powder
Ash 1.5% max.
Quantity 50 g
Percent Purity ≥90%
Infrared Spectrum Authentic
Loss on Drying 10.0% max.
Packaging Plastic bottle
Viscosity 50 to 200mPa·s
Chemical Name or Material Chitosan

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.