missing translation for 'onlineSavingsMsg'
Learn More

Chitosan, MP Biomedicals™

Product Code. 11447901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447901 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447901 Supplier MP Biomedicals Supplier No. 0215059783
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Forms gels with multivalent anions. Chitosan, MP Biomedicals is a cationic marine polymer derived from the shells of crustaceans. It produces clear solutions that dry into strong, clear films. It is a biocompatible, antibacterial and environmentally frien

This chitosan product is a partially de-acetylated chitin formed by the reaction of chitin with concentrated alkali. Chitin is a high molecular weight linear polymer of anhydro-N-acetyl-glucosamine. Chitin occurs in nature as part of the protective coating of some invertebrates, such as marine crustaceans (e.g. crabs). Chitosan polymer is biodegradable, with low toxicity, if any.

Chitosan forms gels with multivalent anions. It produces strong, clear films that have a variety of applications, including: water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.

Specifications

CAS 9012-76-4
Density bulk density: 0.15–0.3g/cm3 (Lit.)
Molecular Formula C56H103N9O39
Quantity 250 g
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
Molecular Weight (g/mol) 1526.464
PubChem CID 71853
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.