missing translation for 'onlineSavingsMsg'
Learn More

Chitosan, TRC

Product Code. 30354208
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354208 100 g 100g
30351585 25 g 25g
30352968 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354208 Supplier Toronto Research Chemicals Supplier No. TRCC315015100G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-76-4
Molecular Formula [C7H15NO4]n
Molecular Weight (g/mol) 1526.5
Synonym 100D-VL, Amidan, Armour-Zen, BC 10, BC 10 (polysaccharide), Batch LO 7319CsU, Biochemica, Biochikol, Biochikol 020PC, Biopolymer L 112, C 3646, C 60M, CG 10, CG 110, ChitoClear TM 850-2, ChitoHeal, Chitolife, Chitopearl 3510, Cytochit, D 06188, DAC 100, Deacetylchitin, Elexa 4, Epakitin, FCM 117, FH 80, FM 200, FM 200 (chitosan), FM 80, FM 80S, Flonac 60M, Flonac C, Flonac C 60M, Flonac C 60M-SEK, Flonac CS, Flonac H, Flonac H 100M, Flonac LV, Flonac N, Flonac NSW-SEK, HC 1, HC 1 (polysaccharide), HEP 40, Haishan KG, Hidagem HCMF, Hiset KW 5, Hydagan DCMF, imica Chitosan
IUPAC Name (2R,3R,4R,5S,6R)-3-amino-5-(((2S,3R,4R,5S,6R)-3-amino-5-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-2,4-diol Polymer
SMILES N[C@@H]1[C@@H](O)[C@H](O[C@H]2[C@H](N)[C@@H](O)[C@H](O[C@H]3[C@H](N)[C@@H](O)[C@H](O)[C@@H](CO)O3)[C@@H](CO)O2)[C@@H](CO)O[C@H]1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C18H35N3O13/c19-7-12(27)14(5(2-23)30-16(7)29)33-18-9(21)13(28)15(6(3-24)32-18)34-17-8(20)11(26)10(25)4(1-22)31-17/h4-18,22-29H,1-3,19-21H2/t4-,5-,6-,7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17+,18+/m1/s1
Chemical Name or Material Chitosan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.