missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol 2-(O-tert-Butyldimethylsilyl)methyl 1-Acetate, TRC

Product Code. 30353658
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353658 Supplier Toronto Research Chemicals Supplier No. TRCC32510010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloramphenicol 2-(O-tert-Butyldimethylsilyl)methyl 1-Acetate
CAS 864529-27-1
Molecular Formula C19H28Cl2N2O6Si
Quantity 10 mg
Synonym N-[(1R,2R)-2-(Acetyloxy)-1-(O-tert-butyldimethylsilyl)methyl-2-(4-nitrophenyl)ethyl]-2,2-dichloro-acetamide, N-[(1R,2R)-2-(Acetyloxy)-1-[[[(1,1-dimethylethyl)dimethylsilyl]oxy]methyl]-2-(4-nitrophenyl)ethyl]-2,2-dichloro-acetamide.
InChI Formula InChI=1S/C19H28Cl2N2O6Si/c1-12(24)29-16(13-7-9-14(10-8-13)23(26)27)15(22-18(25)17(20)21)11-28-30(5,6)19(2,3)4/h7-10,15-17H,11H2,1-6H3,(H,22,25)/t15-,16+/m1/s1
SMILES CC(=O)O[C@H]([C@@H](CO[Si](C)(C)C(C)(C)C)NC(=O)C(Cl)Cl)c1ccc(cc1)[N+](=O)[O-]
IUPAC Name [(1S,2R)-3-[tert-butyl(dimethyl)silyl]oxy-2-[(2,2-dichloroacetyl)amino]-1-(4-nitrophenyl)propyl] acetate
Molecular Weight (g/mol) 479.43
Formula Weight 478.1094
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.