missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol Crystalline MP Biomedicals

Product Code. 11461861 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Unit Size:
1kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461861 1 kg 1kg
11421861 5 g 5g
11451861 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461861 Supplier MP Biomedicals Supplier No. 0219032191
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
Synonym chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Density 1.586 g/mL
Quantity 1 kg
Formula Weight 323.15
Physical Form Crystalline
Chemical Name or Material Chloramphenicol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.