missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chloramphenicol sodium succinate, 98-102%

Product Code. 15539483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15539483 25 g 25g
15529483 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15539483 Supplier Thermo Scientific Chemicals Supplier No. 459530250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 982-57-0
Molecular Formula C15H15Cl2N2NaO8
Molecular Weight (g/mol) 445.18
InChI Key RPLOPBHEZLFENN-HTMVYDOJSA-M
Synonym succinic acid chloramphenicol sodium
PubChem CID 73940259
IUPAC Name sodium;4-[(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propoxy]-4-oxobutanoate
SMILES C1=CC(=CC=C1C(C(COC(=O)CCC(=O)[O-])NC(=O)C(Cl)Cl)O)[N+](=O)[O-].[Na+]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.0 to 102% on dried substance
Quantity 25 g
Formula Weight 445.18
Percent Purity 98 to 102%
Chemical Name or Material Chloramphenicol sodium succinate
compliance-icons
Product Identifier
  • Chloramphenicol sodium succinate
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.