missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol Succinate Sodium (>90%), TRC

Product Code. 30136137
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136137 5 g 5g
30134724 25 g 25g
30004450 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136137 Supplier Toronto Research Chemicals Supplier No. TRCC3250655G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 982-57-0
Molecular Formula C15 H15 Cl2 N2 O8 . Na
Molecular Weight (g/mol) 445.18
Synonym Butanedioic acid 1-[(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] ester sodium salt (1:1), Butanedioic acid mono[(2R,3R)-2-[(dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] ester monosodium salt, Butanedioic acid [R-(R*,R*)]-mono[2-[(dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] ester monosodium salt, Succinic acid α-monoester with D-threo-(-)-2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]acetamide monosodium salt, Betamicetin, Chloramphenicol monosuccinate sodium salt, Chloramphenicol sodium hemisuccinate, Chloramphenicol sodium monosuccinate, Chloramphenicol sodium succinate, Chloramphenicol succinate sodium, Chloramphenicol succinate sodium salt, Chlorocid S, Chloromycetin sodium succinate, Cidocetine, Clorofenicina, Cloromoin, D-Chloramphenicol sodium succinate, Levomycetin sodium succinate, Levomycetin-Nalcein, Mychel-S, Nalecin, Nevimycin, Protophenicol, Sodium chloramphenical monosuccinate, Sodium chloramphenicol 3-succinate, Sodium chloramphenicol succinate, Solnicol
IUPAC Name sodium;4-[(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propoxy]-4-oxobutanoate
SMILES [Na+].O[C@@H]([C@@H](COC(=O)CCC(=O)[O-])NC(=O)C(Cl)Cl)c1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H16Cl2N2O8.Na/c16-14(17)15(24)18-10(7-27-12(22)6-5-11(20)21)13(23)8-1-3-9(4-2-8)19(25)26;/h1-4,10,13-14,23H,5-7H2,(H,18,24)(H,20,21);/q;+1/p-1/t10-,13-;/m1./s1
Formula Weight 444.0103
Chemical Name or Material Chloramphenicol Sodium Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.