missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorazol Black E, pure

Product Code. 10732782 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10732782

Brand: Thermo Scientific Chemicals 404400250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1937-37-7 | C34H25N9Na2O7S2 | 781.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1937-37-7
Molecular Formula C34H25N9Na2O7S2
Molecular Weight (g/mol) 781.73
MDL Number MFCD00066363
InChI Key OLVNAWLXZDRGPL-UHFFFAOYSA-L
PubChem CID 44135447
IUPAC Name disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Absorption Ratio 0.97 to 1
Infrared Spectrum Authentic
Lambda Max 597 to 602nm
Formula Weight 781.73
Grade Pure
Packaging Glass bottle
Color Black
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Chlorazol Black E, Pure
compliance-icons
Product Identifier
  • Chlorazol Black E, certified
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.