missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chlorazol Black E

Brand:  Thermo Scientific™ A13259.18

Quantity 50g

 View more versions of this product

Product Code. 11430803

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1937-37-7
781.73
OLVNAWLXZDRGPL-UHFFFAOYSA-L
44135447
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C34H25N9Na2O7S2
MFCD00066363
Azo Black; C.I. 30235
disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate

Specifications

1937-37-7
MFCD00066363
OLVNAWLXZDRGPL-UHFFFAOYSA-L
disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44135447
50g
C34H25N9Na2O7S2
Azo Black; C.I. 30235
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
781.73
781.73
Chlorazol Black E
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.