missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Digluconate (20% in water), TRC

Product Code. 30012436
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012436 50 g 50g
30005701 1 g 1g
30011299 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012436 Supplier Toronto Research Chemicals Supplier No. TRCC29277050G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C22 H30 Cl2 N10 . 2 C6 H12 O7
Molecular Weight (g/mol) 897.76
Synonym D-Gluconic acid, compd. with N1,N14-bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide (2:1), D-Gluconic acid, compd. with 1,1'-hexamethylenebis[5-(p-chlorophenyl)biguanide] (2:1) (6CI), D-Gluconic acid, compd. with N,N' '-bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide (2:1) (9CI), Gluconic acid, compd. with 1,1'-hexamethylenebis[5-(p-chlorophenyl)biguanide] (2:1), D- (8CI), 2,4,11,13-Tetraazatetradecanediimidamide, N,N' '-bis(4-chlorophenyl)-3,12-diimino-, di-D-gluconate (9CI), Biguanide, 1,1'-hexamethylenebis[5-(p-chlorophenyl)-, di-D-gluconate (8CI), 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] digluconate, 1,6-Bis(4-chlorophenyldiguanino)hexane digluconate, 1,6-Bis(p-chlorophenyldiguanido)hexane digluconate, 1,6-Bis[N5-(p-chlorophenyl)biguanido]hexane digluconate, Abacil, Acclean, Akhdez 3000, Antiseptol, Arlacide G, Bacticlens, Betasept, Bis(p-chlorophenyl)diguanidohexane digluconate, Blue Ribbon Sanitizing Teat Dip, CHG Solution BP, Chloraprep, Chlorhexamed, Chlorhexidine bigluconate, Chlorhexidine di-D-gluconate, Chlorhexidine digluconate, Chlorhexidine gluconate, Corsodyl, Disteryl, Geksikon, Gingisan, Hexana, Hexicon, Hexidine, Hibidil, Hibiscrub, Hibisol, Hibistat, Hibital, Hibitane, Hibitane 5, Hibitane G, Manusan, Maskin, Maskin R, Medihex-4, Oramed, PHiso-Med, Peridex, Peridex (antiseptic), PerioChip, Permachem CL 40, Plac Out, Purit, Rotersept, SY 1007, Secalan, Septeal, Sterilon, Unisept
IUPAC Name 1-(4-chlorophenyl)-3-[N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C22H30Cl2N10.2C6H12O7/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*7-1-2(8)3(9)4(10)5(11)6(12)13/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*2-5,7-11H,1H2,(H,12,13)/t;2*2-,3-,4+,5-/m.11/s1
Formula Weight 896.3198
Percent Purity >95%
Chemical Name or Material Chlorhexidine digluconate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.