missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Dihydrochloride Impurity B, TRC

Product Code. 30007811
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007811 25 mg 25mg
30010544 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007811 Supplier Toronto Research Chemicals Supplier No. TRCC37768025MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308292-89-8
Molecular Formula C16 H26 Cl N9 O
Molecular Weight (g/mol) 395.89
Synonym 2,4,11,13-Tetraazatetradecanamide, 14-[(4-chlorophenyl)amino]-3,12,14-triimino-, Chlorhexidine urea, 1-[[6-[[[(4-Chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]urea
IUPAC Name [N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]urea
SMILES NC(=O)NC(=N)NCCCCCCNC(=N)NC(=N)Nc1ccc(Cl)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H26ClN9O/c17-11-5-7-12(8-6-11)24-15(20)25-13(18)22-9-3-1-2-4-10-23-14(19)26-16(21)27/h5-8H,1-4,9-10H2,(H5,18,20,22,24,25)(H5,19,21,23,26,27)
Formula Weight 395.1949
Percent Purity >95%
Chemical Name or Material Chlorhexidine urea
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.