missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, TRC

Product Code. 30366930
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366930 5 g 5g
30360845 1 g 1g
30352108 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366930 Supplier Toronto Research Chemicals Supplier No. TRCC3350455G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-56-1
Molecular Formula C22 H30 Cl2 N10
Molecular Weight (g/mol) 505.45
Synonym 2,4,11,13-Tetraazatetradecanediimidamide, N,N''-bis(4-chlorophenyl)-3,12-diimino- (9CI), Biguanide, 1,1'-hexamethylenebis[5-(p-chlorophenyl)- (6CI,7CI,8CI), N1,N14-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide, 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide], 1,6-Bis[5-(p-chlorophenyl)biguanidino]hexane, 1,6-Di(N-p-chlorophenylbiguanidino)hexane, Chlorhexamed forte, Chlorhexidine, Chlorohex, Chlorohexidene, Chlorohexidine, Consepsis, Dentosan, Dezin, Eburos, Fimeil, Hexadol, Hexident, Nolvasan, Periogard, Promax, Soretol, Tubulicid
IUPAC Name 1-(4-chlorophenyl)-3-[N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]guanidine
SMILES Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H30Cl2N10/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34)
Formula Weight 504.2032
Percent Purity >95%
Chemical Name or Material Chlorhexidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.