missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloridazon-d5 (phenyl-d5) (C.P. >95%), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Chloridazon phenyl-d5 |
| CAS | 1246818-99-4 |
| Molecular Formula | C10 2H5 H3 Cl N3 O |
| Quantity | 0.005 g |
| Synonym | Chloridazon D5 (phenyl D5) |
| InChI Formula | InChI=1 S/C10H8ClN3O/c11-9-8(12)6-13-14(10(9)15)7-4-2-1-3-5-7/h1-6 H,12H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N2N=CC(=C(Cl)C2=O)N)c([2 H])c1[2 H] |
| IUPAC Name | 5-amino-4-chloro-2-(2,3,4,5,6-pentadeuteriophenyl)pyridazin-3-one |
| Molecular Weight (g/mol) | 226.67 |
| Formula Weight | 226.067 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction