Learn More
Palintest™ Chlorine DPD 1 & 3 Photometer Reagents
Product Code. 11924666
Click to view available options
Range:
0 to 5mg/L
Unit Size:
Pack of 50
Product Code. 11924666
Brand: Palintest™ PM031
Test for chlorine using these tablets that rely on the DPD method developed by Dr. A. T. Palin. Palintest™ Chlorine DPD 1 & 3 Photometer Reagents are very convenient and easy to use.
  • DPD method - standard method for testing for chlorine and other disinfectant residuals.
  • Tablet form for maximum convenience and simplicity of use.
  • Measures free chlorine concentration and total chlorine concentration.
  • Measure color intensities with a Palintest™ photometer.
Specifications
No. of Tests 50
Quantity 50/Pk.
Range 0 to 5mg/L
Sufficient For 50 Tests
Type Chlorine DPD 1 & 3 Test Reagent Tablets