missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Chlorine DPD 2 Test Kit

Product Code. 11983607
Click to view available options
Range:
0 to 5mg/L
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11983607

Brand: Palintest™ PM021

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

No. of Tests 50
Quantity 50 Pack
Range 0 to 5mg/L
Sufficient For 50 Tests
Type Chlorine DPD 2 Test Kit
compliance-icons
Product Identifier
  • X50 TEST CHLORE DPD N°2 0-5PPM
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromide, potassium iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.