missing translation for 'onlineSavingsMsg'
Learn More

Chlormadinone acetate, MedChemExpress

Product Code. 16468606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468606 Supplier Medchem Express Supplier No. HYB1095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chlormadinone acetate is a steroidal progestin, with antiandrogen and antiestrogenic effects.

Chlormadinone acetate is a steroidal progestin, with antiandrogen and antiestrogenic effects.

Specifications

Chemical Name or Material Chlormadinone acetate
CAS 302-22-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H29ClO4
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (24.70 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC([C@@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])C=C(Cl)C4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 404.93
Formula Weight 404.93
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.