missing translation for 'onlineSavingsMsg'
Learn More

Chlormadinone Acetate, TRC

Product Code. 30012427
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012427 10 mg 10mg
30005493 100 mg 100mg
30005277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012427 Supplier Toronto Research Chemicals Supplier No. TRCC34350010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-22-7
Molecular Formula C23 H29 Cl O4
Molecular Weight (g/mol) 404.93
Synonym Pregna-4,6-diene-3,20-dione, 6-chloro-17-hydroxy-, acetate (6CI,7CI,8CI), 17-(Acetyloxy)-6-chloropregna-4,6-diene-3,20-dione, 17-Acetoxy-6-chloro-6-dehydroprogesterone, 17-Acetoxy-6-chloropregna-4,6-diene-3,20-dione, 17α-Acetoxy-6-chloro-4,6-pregnadiene-3,20-dione, 17α-Acetoxy-6-chloro-6,7-dehydroprogesterone, 6-Chloro-17-acetoxy-4,6-pregnadiene-3,20-dione, 6-Chloro-17-hydroxypregna-4,6-diene-3,20-dione acetate, 6-Chloro-17α-acetoxy-4,6-pregnadiene-3,20-dione, 6-Chloro-17α-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, 6-Chloro-17α-hydroxy-δ6-progesterone acetate, 6-Chloro-17α-hydroxypregna-4,6-diene-3,20-dione acetate, 6-Chloro-6,7-dehydro-17-acetoxyprogesterone, 6-Chloro-6-dehydro-17-acetoxyprogesterone, 6-Chloro-6-dehydro-17α-acetoxyprogesterone, 6-Chloro-pregna-4,6-dien-17α-ol-3,20-dione acetate, 6-Chloro-δ4,6-pregnadien-17α-ol-3,20-dione 17-acetate, 6-Chloro-δ6-17-acetoxyprogesterone, 6-Chloro-δ6-[17α]acetoxyprogesterone, 6-Chloro-δ6-dehydro-17-acetoxyprogesterone, 6-Dehydro-6-chloro-17α-acetoxyprogesterone, Ay 13390-6, Bellissima, Bovisynchron, CAP, Cero, Chlormadinon acetate, Chlormadinone acetate, Chronosyn, Clordion, Cyclonorm, Fertiletten, Gestafortin, Lormin, Luteran, Lutinyl, Lutoral, Lutoral (Syntex), Matrol, Menstridyl, NSC 92338, Natrol, Normenon, Prostal, RS 1280, Retex, Skedule TM, Synchrosyn, Synchrosyn P, Traslan, Verton
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6-chloro-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H29ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,16-18H,5-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1
Formula Weight 404.1754
Percent Purity >95%
Chemical Name or Material Chlormadinone acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.