missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Chlormezanone
Description
Chlormezanone is a anxiolytic and skeletal muscle relaxant that acts at the benzodiazepine site of GABAA receptors.
Specifications
Specifications
| Chemical Name or Material | 2-(4-Chlorophenyl)tetrahydro-3-methyl-4H-1,3-thiazin-4-one-1,1-dioxide |
| CAS | 80-77-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | GABA-A Receptor Benzodiazepines |
| Molecular Formula | C11H12ClNO3S |
| Purity | 0.99 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?