missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) Dimer, TRC

Product Code. 30350954
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350954 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350954 Supplier Toronto Research Chemicals Supplier No. TRCC293350500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloro(1,5-cyclooctadiene)rhodium(I) Dimer
CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Quantity 500 mg
Synonym (1,5-Cyclooctadiene)Rhodium Chloride Dimer, (1,5-Cyclooctadiene) Rhodium(I) Chloride Dimer, (eta4-1,5-Cyclooctadiene)Rhodium Chloride Dimer, Bis((1,5-cyclooctadiene)(Chloro)Rhodium), Bis((μ-chloro)(eta4-1,5-Cyclooctadiene)Rhodium), Bis(1,5-cyclooctadiene)Dirhodium Dichloride, Bis(chloro(1,5-cyclooctadiene)Rhodium), Bis(chloro(eta4-1,5-cyclooctadiene)rhodium), Bis(cyclooctadiene)Dichlorodirhodium, Bis(cyclooctadienerhodium Chloride), Chlororhodium(1,5-cyclooctadiene) Dimer, Cyclooctadiene Rhodium Chloride Dimer, Di-μ-Chlorobis(1,5-cyclooctadiene)Dirhodium, Di-μ-chlorobis(cyclooctadiene) Dirhodium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)dirhodium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)Dirhodium(I), Di-μ-chlorobis[(1,2,5,6-eta)-1,5-cyclooctadiene]Dirhodium, Di-μ-chlorobis[(1,5-cyclooctadiene)Rhodium], Di-μ2-chlorobis(1,5-cyclooctadiene)Dirhodium, Dichlorobis(1,5-cyclooctadiene)Dirhodium, Dichlorobis(1,5-cyclooctadiene)Rhodium(I), Dichlorobis(cyclooctadiene)Dirhodium, Rhodium 1,5-Cyclooctadiene chloride dimer, μ-Chloro(1,5-cyclooctadiene)Rhodium(I) Dimer, Bis(1,5-cyclooctadienedirhodium(I) Chloride)
InChI Formula InChI=1S/2C8H12.2ClH.2Rh/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;
SMILES Cl[Rh].Cl[Rh].C1C\C=C/CC\C=C/1.C2C\C=C/CC\C=C/2
IUPAC Name chlororhodium;(1Z,5Z)-cycloocta-1,5-diene
Molecular Weight (g/mol) 493.08
Formula Weight 491.9365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.