missing translation for 'onlineSavingsMsg'
Learn More

Chloro Chlorthalidone, TRC

Product Code. 30135436
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135436 25 mg 25mg
30132935 100 mg 100mg
30008908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135436 Supplier Toronto Research Chemicals Supplier No. TRCC42752025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16289-13-7
Molecular Formula C14 H9 Cl2 N O2
Molecular Weight (g/mol) 294.13
Synonym 1H-Isoindol-1-one, 3-(3,4-dichlorophenyl)-2,3-dihydro-3-hydroxy-, Phthalimidine, 3-(3,4-dichlorophenyl)-3-hydroxy- (7CI,8CI), (3RS)-3-(3,4-Dichlorophenyl)-3-hydroxy-2,3-dihydro-1H-isoindol-1-one, 3-(3,4-Dichlorophenyl)-3-hydroxy-isoindolin-1-one
IUPAC Name 3-(3,4-dichlorophenyl)-3-hydroxy-2H-isoindol-1-one
SMILES OC1(NC(=O)c2ccccc12)c3ccc(Cl)c(Cl)c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H9Cl2NO2/c15-11-6-5-8(7-12(11)16)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)
Formula Weight 293.001
Percent Purity >95%
Chemical Name or Material chloro-Chlorthalidone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.