missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde diethyl acetal, 98%

Product Code. 15468869
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15468869

Brand: Thermo Scientific Alfa Aesar L07661.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-62-5 | C6H13ClO2 | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC

Specifications

Melting Point -45°C
Density 1.018
Boiling Point 156°C to 157°C
Flash Point 50°C (122°F)
Linear Formula ClCH2CH(OCH2CH3)2
Refractive Index 1.416
Quantity 100 g
UN Number UN1993
Beilstein 969254
Sensitivity Moisture sensitive
Formula Weight 152.62
Percent Purity 98%
Chemical Name or Material Chloroacetaldehyde diethyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroacetaldehyde diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Chloroacetaldehyde diethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.