missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde diethyl acetal, 99%, Thermo Scientific™

Product Code. 10743831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743831 5 mL Glass bottle 5mL
10345740 100 mL Glass bottle 100mL
10497670 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10743831 Supplier Thermo Scientific Acros Supplier No. 108490050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC

Specifications

Melting Point -45.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 157.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClCH2CH(OC2H5)2
Refractive Index 1.4150 to 1.4170
Quantity 5 mL
Beilstein 01, 611
Fieser 05,107
Specific Gravity 1.01
Solubility Information Solubility in water: 1.8g/L (20 c).
Formula Weight 152.62
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Chloroacetaldehyde diethyl acetal, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroacetaldehyde diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.