missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde Diethyl Acetal, TRC

Product Code. 30356726
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356726 Supplier Toronto Research Chemicals Supplier No. TRCC2927851G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroacetaldehyde Diethyl Acetal
CAS 621-62-5
Molecular Formula C6H13ClO2
Quantity 1 g
Synonym 1-Diethoxy-2-chloro-1 Ethane, Chloro-acetaldehyde Diethyl Acetal, 1-Chloro-2,2-diethoxyethane, 2-Chloro-1 1-Diethoxyethane, 2-Chloroacetaldehyde Diethyl Acetal, Chloroacetaldehyde Diethyl Acetal, Chloroethanal Diethyl Acetal, Diethoxyethyl Chloride, Monochloroacetaldehyde Diethyl Acetal, NSC 8436
InChI Formula InChI=1S/C6H13ClO2/c1-3-8-6(5-7)9-4-2/h6H,3-5H2,1-2H3
SMILES CCOC(CCl)OCC
IUPAC Name 2-chloro-1,1-diethoxyethane
Molecular Weight (g/mol) 152.62
Formula Weight 152.0604
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.