missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamidine hydrochloride, 96%

Product Code. 11492597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492597 5 g 5g
11482597 1 g 1g
11402607 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492597 Supplier Thermo Scientific Alfa Aesar Supplier No. A14835.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10300-69-3 | C2H6Cl2N2 | 128.98 g/mol

Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10300-69-3
Molecular Formula C2H6Cl2N2
Molecular Weight (g/mol) 128.98
MDL Number MFCD00053013
InChI Key GUPOZVHRTJYZCX-UHFFFAOYSA-N
Synonym chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
PubChem CID 2776957
IUPAC Name 2-chloroethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 5 g
UN Number UN3261
Beilstein 3678354
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 129
Percent Purity 96%
Chemical Name or Material Chloroacetamidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.