missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic acid, ACS, 99%

Product Code. 11368878
Change view
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
Unit Size:
100g
2000g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368878 500 g 500g
11348878 100 g 100g
11358878 4 x 500 g 2000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368878 Supplier Thermo Scientific Alfa Aesar Supplier No. 041724.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 79-11-8 | C2H3ClO2 | 94.494 g/mol

Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.494
MDL Number MFCD00002683
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Melting Point 60°C to 63°C
Density 1.404
Boiling Point 189°C
Flash Point 126°C (258°F)
Linear Formula CH2CO2H
Quantity 500 g
UN Number UN1751
Beilstein 605438
Merck Index 14,2112
Solubility Information Soluble in methanol,acetone,diethyl ether,benzene,chloroform and ethanol.
Formula Weight 94.5
Percent Purity 99%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Chloroacetic Acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.