missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 199.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass Bottle |
| Linear Formula | CH2ClCOONa |
| Quantity | 2.5 kg |
| Merck Index | 15, 2112 |
| Solubility Information | Solubility in water: soluble in water: 440g/L (20°C).. Other solubilities: slightly soluble in methanol., insoluble in acetone, benzene, ether, ccl4. |
| Formula Weight | 116.48 |
| Show More |
Safety and Handling
Produktidentifikator
- Chloroacetic acid sodium salt
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H301-Toxic if swallowed.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?