missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic Acid, TRC

Product Code. 30133774
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133774 10 g 10g
30041001 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133774 Supplier Toronto Research Chemicals Supplier No. TRCC36336510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2 H3 Cl O2
Molecular Weight (g/mol) 94.5
Synonym Acetic acid, chloro- (8CI,9CI), 2-Chloroacetic acid, Chloracetic acid, Chloroacetic acid, Chloroethanoic acid, MCA, MCAA, MKhUK, Monochloroacetic acid, Monochloroethanoic acid, NSC 142, NSC 42970, α-Chloroacetic acid
IUPAC Name 2-chloroacetic acid
SMILES OC(=O)CCl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)
Formula Weight 93.9822
Percent Purity >95%
Chemical Name or Material Chloroacetic acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.