missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic anhydride, 97%

Product Code. 11467886
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467886 500 g 500g
11447886 25 g 25g
11457886 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467886 Supplier Thermo Scientific Alfa Aesar Supplier No. A14015.36
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-88-8 | C4H4Cl2O3 | 170.97 g/mol

Chloroacetic anhydride has been used in the synthesis of 3,3?-bis(sulfonato)-4,4?-bis(chloroacetamido)azobenzene (BSBCA), a water-soluble, thiol-reactive and photo-switchable cross-linker, D,L-7-azatryptophan, 2-methyl-[3,4-di-O-acetyl-6-O-(chloroacetyl)-1,2-dideoxy-α-D-glucopyrano]-[2?,1?:4,5]-2-oxazoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetic anhydride has been used in the synthesis of 3,3′-bis(sulfonato)-4,4′-bis(chloroacetamido)azobenzene (BSBCA), a water-soluble, thiol-reactive and photo-switchable cross-linker, D,L-7-azatryptophan, 2-methyl-[3,4-di-O-acetyl-6-O-(chloroacetyl)-1,2-dideoxy-α-D-glucopyrano]-[2′,1′:4,5]-2-oxazoline.

Solubility
Freely soluble in ether, chloroform. Slightly soluble in benzene. Insoluble in cold ligroin

Notes
Moisture sensitive. Incompatible with oxidizing agent, bases, amines, alcohols, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-88-8
Molecular Formula C4H4Cl2O3
Molecular Weight (g/mol) 170.97
MDL Number MFCD00000929
InChI Key PNVPNXKRAUBJGW-UHFFFAOYSA-N
Synonym chloroacetic anhydride, chloracetic anhydride, monochloroacetic acid anhydride, 2-chloroacetic anhydride, chloroacetic acid anhydride, chloroacetyl anhydride, acetic acid, chloro-, anhydride, unii-8q7txh7r5f, monochloroacetic anhydride, 8q7txh7r5f
PubChem CID 10946
IUPAC Name (2-chloroacetyl) 2-chloroacetate
SMILES ClCC(=O)OC(=O)CCl

Specifications

Melting Point 56°C to 59°C
Boiling Point 110°C to 112°C (10 mmHg)
Flash Point 143°C (289°F)
Quantity 500 g
UN Number UN3261
Beilstein 774533
Sensitivity Moisture sensitive
Merck Index 14,2113
Solubility Information Freely soluble in ether, chloroform; Slightly soluble in benzene; Insoluble in cold ligroin
Formula Weight 170.98
Percent Purity 97%
Chemical Name or Material Chloroacetic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.